4-Acetoxy-2-azetidinone 97%

Supplier: Ambeed
Warning

A197756-100G A197756-10G A197756-250MG A197756-1G A197756-5G
103816-964EA 23 USD
103816-964 103847-586 77012-548 77012-550 77012-552 77085-598 77085-600
4-Acetoxy-2-azetidinone 97%
4-Acetoxy-2-azetidinone

Precaución: for Research Use Only

Formula: C₅H₇NO₃
Boiling Pt: 296.5 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 133.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010593
CAS Number: 28562-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR