4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-Acetamido-2,2,6,6-Tetramethyl-N-Piperidinyloxy
A407910-1G
A407910-5G
A407910-100G
A407910-10G
A407910-25G
103827-168EA
12
USD
103827-168
103838-210
76841-704
76841-706
77291-264
77291-266
77307-118
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) 98%
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO)
Precaución: for Research Use Only
Formula:
C₁₁H₂₁N₂O₂ MW: 213.3 g/mol Boiling Pt: 368.8 °C (760 mm Hg) Melting Pt: 143…145 °C Flash Pt: 176.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043593 CAS Number: 14691-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...