3-(Morpholino)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Morpholino)phenylboronic acid Pinacol ester, 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 2-[3-(Morpholino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A290145-100MG A290145-1G A290145-5G
103813-862EA 14 USD
103813-862 103835-004 77114-250 77305-576 77305-578
3-(Morpholino)phenylboronic acid pinacol ester 98%
3-(Morpholino)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₃
MW: 289.1777 g/mol
Boiling Pt: 431.5 °C (760 mm Hg)
Melting Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD03412097
CAS Number: 852227-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR