4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride

A517625-25G A517625-100G A517625-500G A517625-5G
103828-476EA 5 USD
103828-476 103839-580 76843-576 76979-992 76979-994 77307-692
4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride 98%
4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride

Precaución: For research use only

Formula: C₁₃H₁₈N₂O₂
MW: 307.22 g/mol
Melting Pt: 305…307 °C
Storage Temperature: Ambient
MDL Number: MFCD07772867
CAS Number: 106261-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR