Erlotinib 95%

Supplier: Ambeed
Warning

Synonyms: 4-[(3-Ethynylphenyl)amino]-6,7-bis(2-methoxyethoxy)quinazoline

A121900-1MG A121900-25G A121900-1G A121900-5G A121900-250MG A121900-100G
103800-152EA 15 USD
103800-152 76797-418 76797-420 77312-280 77312-282 77599-752 77666-750
Erlotinib 95%
Erlotinib

Precaución: for Research Use Only

Formula: C₂₂H₂₃N₃O₄
MW: 393.4357 g/mol
Boiling Pt: 553.6 °C (760 mm Hg)
Melting Pt: 223…228 °C
Flash Pt: 288.6 °C
Storage Temperature: Freezer
MDL Number: MFCD08063588
CAS Number: 183321-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR