4,8-Dibromobenzo[1,2-c:4,5-c']bis([1,2,5]thiadiazole) 95%
Supplier: Ambeed
|
Warning
|
A169921-100MG
A169921-250MG
A169921-1G
103806-428EA
285
USD
103806-428
77011-554
77011-556
77301-012
4,8-Dibromobenzo[1,2-c:4,5-c']bis([1,2,5]thiadiazole) 95%
4,8-Dibromobenzo[1,2-c:4,5-c']bis([1,2,5]thiadiazole)
Precaución: for Research Use Only
Formula:
C₆Br₂N₄S₂ MW: 352.03 g/mol Boiling Pt: 469.3±55.0 °C (760 mm Hg) Flash Pt: 237.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD24539447 CAS Number: 165617-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...