4,7-Dimethoxy-1,10-phenanthroline 97%

Supplier: Ambeed
Warning

A225833-100MG A225833-250MG A225833-1G A225833-25G A225833-5G A225833-100G
103819-204EA 12 USD
103819-204 103850-106 76786-122 76786-124 77088-820 77088-822 77088-824 77600-810
4,7-Dimethoxy-1,10-phenanthroline 97%
4,7-Dimethoxy-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₂
MW: 240.2573 g/mol
Melting Pt: 197…212 °C
Flash Pt: 135.888 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233883
CAS Number: 92149-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR