4,7-Dichloroquinoline 97%

Supplier: Ambeed
Warning

A253994-25G A253994-100G A253994-10G A253994-500G A253994-1000G
103851-972EA 6 USD
103851-972 77091-410 77262-074 77262-076 77262-078 77746-408
4,7-Dichloroquinoline 97%
4,7-Dichloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00006774
CAS Number: 86-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR