4,7-Dichloro-1,10-phenanthroline 97%

Supplier: Ambeed
Danger

A380045-1G A380045-250MG A380045-25G A380045-100MG
103826-662EA 13 USD
103826-662 103837-698 76841-022 76989-970 76989-972 77684-095
4,7-Dichloro-1,10-phenanthroline 97%
4,7-Dichloro-1,10-phenanthroline

Precaución: For research use only.

Formula: C₁₂H₆Cl₂N₂
MW: 249.09 g/mol
Boiling Pt: 395.9 °C (760 mm Hg)
Melting Pt: 243…247 °C
Flash Pt: 225.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233882
CAS Number: 5394-23-0
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR