4,7-Dibromo-2,1,3-benzoxadiazole 97%

Supplier: Ambeed
Warning

Synonyms: 4,7-Dibromobenzo[c][1,2,5]oxadiazole, 4,7-Dibromobenzofurazan

A165411-100MG A165411-10G A165411-250MG A165411-25G A165411-1G A165411-5G
103805-966EA 65 USD
103805-966 103844-220 76812-972 76812-974 76812-976 77300-786 77320-424 77320-426
4,7-Dibromo-2,1,3-benzoxadiazole 97%
4,7-Dibromo-2,1,3-benzoxadiazole

Precaución: for Research Use Only

Formula: C₆H₂Br₂N₂O
MW: 277.90 g/mol
Boiling Pt: 312.4 °C (760 mm Hg)
Flash Pt: 142.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02114623
CAS Number: 54286-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR