4,7-Dibromo-2,1,3-benzothiadiazole 98%

Supplier: Ambeed
Danger

A171470-100G A171470-10G A171470-1G A171470-25G A171470-5G
103806-636EA 10 USD
103806-636 103844-910 76998-218 76998-220 77082-070 77082-072 77082-074
4,7-Dibromo-2,1,3-benzothiadiazole 98%
4,7-Dibromo-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₆H₂Br₂N₂S
MW: 293.97 g/mol
Melting Pt: 186…190 °C
Flash Pt: 151.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00658844
CAS Number: 15155-41-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR