4,7-Dibromo-1,10-phenanthroline hydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,7-Dibromo-1,10-phenanthroline
A133976-100MG
A133976-250MG
A133976-1G
A133976-5G
103801-942EA
58
USD
103801-942
77162-006
77162-008
77298-670
77298-672
4,7-Dibromo-1,10-phenanthroline hydrate 97%
4,7-Dibromo-1,10-phenanthroline hydrate
Precaución: for Research Use Only
Formula:
C₁₂H₆Br₂N₂·xH₂O MW: 338.00 g/mol Melting Pt: 236 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD25562940 CAS Number: 156492-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...