4,7-Diazaspiro[2.5]octane dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
A164210-100MG
A164210-1G
103805-832EA
176
USD
103805-832
103844-092
76973-714
77320-246
4,7-Diazaspiro[2.5]octane dihydrochloride 97%
4,7-Diazaspiro[2.5]octane dihydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₄Cl₂N₂ MW: 185.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11506235 CAS Number: 145122-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...