4,7-Di(2-thienyl)-2,1,3-benzothiadiazole 98%

Supplier: Ambeed
Warning

Synonyms: 4,7-Di(thiophen-2-yl)benzo[c][1,2,5]thiadiazole

A306241-100MG A306241-250MG A306241-1G
103814-456EA 75 USD
103814-456 103835-624 76988-874 76988-876 77305-932
4,7-Di(2-thienyl)-2,1,3-benzothiadiazole 98%
4,7-Di(2-thienyl)-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₁₄H₈N₂S₃
MW: 300.41 g/mol
Boiling Pt: 482.1 °C (760 mm Hg)
Melting Pt: 128…129 °C
Flash Pt: 235.4 °C
Storage Temperature: Ambient
MDL Number: MFCD18252231
CAS Number: 165190-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR