4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole 95%

Supplier: Ambeed
Warning

Synonyms: 2,1,3-Benzothiadiazole-4,7-diboronic Acid Bis(pinacol) Ester, 4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]thiadiazole

A140865-250MG A140865-25G A140865-5G A140865-1G
103802-910EA 27 USD
103802-910 76871-202 76871-204 77299-166 77316-030
4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole 95%
4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₁₈H₂₆B₂N₂O₄S
MW: 388.10 g/mol
Boiling Pt: 482.6±30.0 °C (760 mm Hg)
Melting Pt: 208.5…209.5 °C
Flash Pt: 245.7 °C
Storage Temperature: Ambient
MDL Number: MFCD11656086
CAS Number: 934365-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR