4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%

Supplier: Ambeed
Warning

A258790-100MG A258790-10G A258790-250MG A258790-1G A258790-5G
103812-400EA 49 USD
103812-400 103852-278 77289-164 77289-166 77289-168 77304-752 77304-754
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)

Precaución: for Research Use Only

Formula: C₁₈H₃₆N₂O₆
Boiling Pt: 513.1 °C (760 mm Hg)
Melting Pt: 68…72 °C
Flash Pt: 144.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00005111
CAS Number: 23978-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR