Bis-PEG4-acid 97%

Supplier: Ambeed
Warning

A169076-1G A169076-100MG A169076-250MG A169076-25G
103806-348EA 85 USD
103806-348 103844-604 77081-676 77138-362 77138-364 77685-368
Bis-PEG4-acid 97%
Bis-PEG4-acid

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₈
Storage Temperature: Ambient
MDL Number: MFCD22201545
CAS Number: 31127-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR