4,6-Dimethylpyrimidin-5-ol 97%

Supplier: Ambeed
Warning

A362259-100MG A362259-250MG A362259-1G
103826-308EA 221 USD
103826-308 103837-322 76773-352 76773-354 77306-732
4,6-Dimethylpyrimidin-5-ol 97%
4,6-Dimethylpyrimidin-5-ol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 237.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00466666
CAS Number: 70345-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR