3-Amino-4,6-dimethylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5A-DMP, 4,6-Dimethylpyridin-3-amine

A112572-10G A112572-250MG A112572-5G A112572-1G
103798-040EA 46 USD
103798-040 103823-878 77270-974 77270-976 77297-190 77310-686
3-Amino-4,6-dimethylpyridine 97%
3-Amino-4,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
Boiling Pt: 257.1 °C (760 mm Hg)
Melting Pt: 47…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07439969
CAS Number: 1193-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR