4,6-Dimethyldibenzothiophene 95%

Supplier: Ambeed
Warning

A174747-5G A174747-250MG
103806-972EA 13 USD
103806-972 103845-272 77082-576 77286-978
4,6-Dimethyldibenzothiophene 95%
4,6-Dimethyldibenzothiophene

Precaución: for Research Use Only

Formula: C₁₄H₁₂S
MW: 212.31 g/mol
Boiling Pt: 364.9 °C (760 mm Hg)
Melting Pt: 153…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00216264
CAS Number: 1207-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR