4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A104579-100G A104579-10G A104579-1G A104579-25G A104579-5G
103822-454EA 6 USD
103822-454 77107-418 77107-420 77107-422 77107-424 77283-252
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 97%
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 285…287 °C
Storage Temperature: Ambient
MDL Number: MFCD00006269
CAS Number: 769-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR