4,6-Dimethyl-2-methylsulfonylpyrimidine 98%

Supplier: Ambeed
Warning

A692431-5G A692431-100G A692431-1G A692431-25G
103840-590EA 9 USD
103840-590 76844-812 77280-772 77280-774 77308-132
4,6-Dimethyl-2-methylsulfonylpyrimidine 98%
4,6-Dimethyl-2-methylsulfonylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Boiling Pt: 358.3 °C (760 mm Hg)
Melting Pt: 84…86 °C
Flash Pt: 170.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03788286
CAS Number: 35144-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR