4,6-Dimethoxypyrimidin-5-amine 95%
Supplier: Ambeed
|
Warning
|
A257619-10G
A257619-250MG
A257619-1G
A257619-5G
103812-324EA
16
USD
103812-324
103852-192
76786-910
76786-912
77304-714
77304-716
4,6-Dimethoxypyrimidin-5-amine 95%
4,6-Dimethoxypyrimidin-5-amine
Precaución: for Research Use Only
Formula:
C₆H₉N₃O₂ Melting Pt: 95…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00194013 CAS Number: 15846-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...