4,6-Dihydroxypyrimidine 95%

Supplier: Ambeed
Warning

A394800-500G A394800-25G A394800-100G
103810-266EA 73 USD
103810-266 76817-820 77307-036 77672-952
4,6-Dihydroxypyrimidine 95%
4,6-Dihydroxypyrimidine

Precaución: for Research Use Only

Formula: C₄H₄N₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00016733
CAS Number: 1193-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR