4,6-Difluoroisatin 95%

Supplier: Ambeed
Warning

Synonyms: 4,6-Difluoro-1H-indole-2,3-dione

A156520-10G A156520-1G A156520-5G
103804-898EA 6 USD
103804-898 103843-098 76997-552 77318-886 77318-888
4,6-Difluoroisatin 95%
4,6-Difluoroisatin

Precaución: for Research Use Only

Formula: C₈H₃F₂NO₂
MW: 183.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03618552
CAS Number: 126674-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR