4,6-Dichloropyrimidine-5-carbonitrile 97%

Supplier: Ambeed
Warning

A424278-10G A424278-1G A424278-25G A424278-5G
103827-444EA 23 USD
103827-444 103838-494 76789-168 76842-138 76842-140 76842-142
4,6-Dichloropyrimidine-5-carbonitrile 97%
4,6-Dichloropyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅HCl₂N₃
Flash Pt: 156.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08460393
CAS Number: 5305-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR