4,6-Dichloropyrimidine-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A224019-100G A224019-10G A224019-1G A224019-25G A224019-5G
103819-078EA 7 USD
103819-078 103849-966 76987-116 76987-118 77088-596 77088-598 77088-600
4,6-Dichloropyrimidine-5-carbaldehyde 97%
4,6-Dichloropyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O
MW: 175.99 g/mol
Boiling Pt: 286.1 °C (760 mm Hg)
Melting Pt: 70…72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02257701
CAS Number: 5305-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR