4,6-Dichloropyrimidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A155513-50MG A155513-250MG A155513-100MG
103804-774EA 609 USD
103804-774 103842-970 77028-354 77871-774 77871-773
4,6-Dichloropyrimidine-2-carboxylic acid 95%

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
Boiling Pt: 424 °C (760 mm Hg)
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD09834770
CAS Number: 684220-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR