4,6-Dichloropyrimidine-2-carbonitrile 97%

Supplier: Ambeed
Warning

A183916-100MG A183916-250MG A183916-5G
103815-684EA 83 USD
103815-684 103846-246 77138-814 77138-816 77746-191
4,6-Dichloropyrimidine-2-carbonitrile 97%
4,6-Dichloropyrimidine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅HCl₂N₃
MW: 173.98 g/mol
Boiling Pt: 327.6±45.0 °C (760 mm Hg)
Flash Pt: 151.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09607724
CAS Number: 1451391-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR