4,6-Dichloropyrimidin-5-ol 96%

Supplier: Ambeed
Warning

A144928-100MG A144928-250MG A144928-5G A144928-1G
103803-470EA 34 USD
103803-470 103841-548 76984-414 76984-416 77299-430 77316-806
4,6-Dichloropyrimidin-5-ol 96%
4,6-Dichloropyrimidin-5-ol

Precaución: for Research Use Only

Formula: C₄H₂Cl₂N₂O
MW: 164.97 g/mol
Boiling Pt: 242.2±35.0 °C (760 mm Hg)
Flash Pt: 100.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17392595
CAS Number: 425394-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR