4,6-Dichloropyrimidin-2-ol 95%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dichloropyrimidin-2(1H)-one

A295175-100MG A295175-250MG
103814-046EA 311 USD
103814-046 76988-704 76988-706
4,6-Dichloropyrimidin-2-ol 95%
4,6-Dichloropyrimidin-2-ol

Precaución: for Research Use Only

Formula: C₄H₂Cl₂N₂O
MW: 164.977 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14702755
CAS Number: 6297-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR