4,6-Dichloropyridine-2,3-diamine 95%

Supplier: Ambeed
Warning

A156973-100MG A156973-250MG A156973-5G
103804-948EA 82 USD
103804-948 103843-150 77150-688 77150-690 77300-226
4,6-Dichloropyridine-2,3-diamine 95%
4,6-Dichloropyridine-2,3-diamine

Precaución: for Research Use Only

Formula: C₅H₅Cl₂N₃
MW: 178.02 g/mol
Melting Pt: 188…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13181778
CAS Number: 24484-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR