4,6-Dichloro-N-methylpyrimidin-2-amine 95%
Supplier: Ambeed
|
Warning
|
A105522-1G
A105522-5G
A105522-250MG
103796-894EA
69
USD
103796-894
103822-628
77309-496
77309-498
77610-828
4,6-Dichloro-N-methylpyrimidin-2-amine 95%
4,6-Dichloro-N-methylpyrimidin-2-amine
Precaución: for Research Use Only
Formula:
C₅H₅Cl₂N₃ Melting Pt: 160…164 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02091109 CAS Number: 10397-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...