4,6-Dichloronicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4,6-dichloropyridine-3-carboxylic acid

A149439-100G A149439-10G A149439-500G A149439-1G A149439-25G A149439-5G A149439-1000G
103804-042EA 5 USD
103804-042 103842-160 76812-132 76812-134 76812-136 77317-658 77317-660 77317-662 77671-104
4,6-Dichloronicotinic acid 98%
4,6-Dichloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MW: 192.00 g/mol
Melting Pt: 158…160 °C
Flash Pt: 157.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00234146
CAS Number: 73027-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR