4,6-Dichloronicotinamide 98%

Supplier: Ambeed
Warning

A138142-10G A138142-250MG A138142-25G A138142-5G
103802-564EA 15 USD
103802-564 77285-612 77285-614 77298-964 77315-586
4,6-Dichloronicotinamide 98%
4,6-Dichloronicotinamide

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O
MW: 191.0200 g/mol
Boiling Pt: 302.9 °C (760 mm Hg)
Melting Pt: 153…155 °C
Flash Pt: 137 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01646073
CAS Number: 70593-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR