4,6-Dichloropyridine-3-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dichloronicotinaldehyde

A433858-1G A433858-25G A433858-5G A433858-100G A433858-100MG A433858-10G A433858-250MG
103827-564EA 18 USD
103827-564 103838-632 76842-340 76842-342 76842-344 76877-286 76877-288 76877-290 76877-292
4,6-Dichloropyridine-3-carbaldehyde 97%
4,6-Dichloropyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12025485
CAS Number: 1060811-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR