4,6-Dichloro-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dichloroindole-2-carboxylic acid

A213055-1G A213055-5G A213055-100MG A213055-250MG
103818-258EA 44 USD
103818-258 103848-960 77087-440 77087-442 77261-106 77261-108
4,6-Dichloro-1H-indole-2-carboxylic acid 95%
4,6-Dichloro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅Cl₂NO₂
MW: 230.044 g/mol
Boiling Pt: 476.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00209870
CAS Number: 101861-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR