4,6-Dichloro-5-pyrimidinecarboxylic acid 95%

Supplier: Ambeed
Warning

A183664-1G A183664-5G A183664-10G
103815-656EA 6 USD
103815-656 103846-220 77083-824 77083-826 77287-350
4,6-Dichloro-5-pyrimidinecarboxylic acid 95%
4,6-Dichloro-5-pyrimidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
MW: 192.99 g/mol
Boiling Pt: 331.1 °C (760 mm Hg)
Flash Pt: 161 °C
Storage Temperature: Freezer
MDL Number: MFCD09909611
CAS Number: 87600-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR