4,6-Dichloro-5-nitropyrimidine 98%

Supplier: Ambeed
Warning

A517593-100G A517593-25G A517593-5G
103839-578EA 10 USD
103839-578 76830-342 76843-572 76843-574
4,6-Dichloro-5-nitropyrimidine 98%
4,6-Dichloro-5-nitropyrimidine

Precaución: for Research Use Only

Formula: C₄HCl₂N₃O₂
MW: 193.98 g/mol
Boiling Pt: 325.8 °C (760 mm Hg)
Melting Pt: 100…103 °C
Flash Pt: 150.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006107
CAS Number: 4316-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR