4,6-Dichloro-5-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A192394-100G A192394-10G A192394-500G A192394-5G
103847-042EA 5 USD
103847-042 76986-140 76986-142 76986-144 77084-912
4,6-Dichloro-5-methoxypyrimidine 98%
4,6-Dichloro-5-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂O
MW: 179.01 g/mol
Boiling Pt: 257.8 °C (760 mm Hg)
Melting Pt: 66…68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00087680
CAS Number: 5018-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR