4,6-Dichloro-5-iodopyrimidine 98%

Supplier: Ambeed
Warning

A213007-100MG A213007-250MG A213007-1G
103818-256EA 23 USD
103818-256 103848-956 77030-558 77030-560 77087-436
4,6-Dichloro-5-iodopyrimidine 98%
4,6-Dichloro-5-iodopyrimidine

Precaución: for Research Use Only

Formula: C₄HCl₂IN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18072689
CAS Number: 1137576-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR