4,6-Dichloro-5-fluoropyrimidine 98%

Supplier: Ambeed
Warning

A120053-100G A120053-10G A120053-25G A120053-1G A120053-5G
103799-874EA 5 USD
103799-874 77094-422 77094-424 77094-426 77311-986 77311-988
4,6-Dichloro-5-fluoropyrimidine 98%
4,6-Dichloro-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄HCl₂FN₂
MW: 166.97 g/mol
Boiling Pt: 227 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08056331
CAS Number: 213265-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR