4,6-Dichloro-3-methyl-1H-pyrazolo[4,3-c]pyridine 95%

Supplier: Ambeed
Warning

A219383-100MG A219383-250MG
103818-786EA 100 USD
103818-786 76987-012 76987-014
4,6-Dichloro-3-methyl-1H-pyrazolo[4,3-c]pyridine 95%
4,6-Dichloro-3-methyl-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅Cl₂N₃
MW: 202.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257115
CAS Number: 120422-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR