Fenclorim 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,6-Dichloro-2-phenylpyrimidine
A125431-100G
A125431-10G
A125431-25G
76971-844EA
46.68
USD
76971-844
76971-846
76971-848
Fenclorim 97%
Fenclorim
Precaución: for Research Use Only
Formula:
C₁₀H₆Cl₂N₂ Boiling Pt: 235.5 °C (760 mm Hg) Melting Pt: 92…93 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00754193 CAS Number: 3740-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...