4,6-Dichloro-2-(methylsulfonyl)pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dichloro-2-(methanesulphonyl)pyrimidine, 4,6-Dichloro-2-methylsulphonylpyrimidine

A119908-100G A119908-10G A119908-25G A119908-500G A119908-1G A119908-5G
103799-856EA 5 USD
103799-856 76971-436 76971-438 76971-440 76971-442 77297-724 77311-964
4,6-Dichloro-2-(methylsulfonyl)pyrimidine 98%
4,6-Dichloro-2-(methylsulfonyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂O₂S
MW: 227.06 g/mol
Boiling Pt: 402.3 °C (760 mm Hg)
Melting Pt: 117…118 °C
Flash Pt: 197.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00672147
CAS Number: 4489-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR