4,6-Dichloro-2-methylpyrido[3,4-d]pyrimidine 97%

Supplier: Ambeed
Warning

A101005-100MG A101005-250MG A101005-1G
103795-748EA 191 USD
103795-748 103821-440 76821-220 76821-222 77296-946
4,6-Dichloro-2-methylpyrido[3,4-d]pyrimidine 97%
4,6-Dichloro-2-methylpyrido[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N₃
MW: 214.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044721
CAS Number: 1029720-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR