4,6-Dichloro-2-cyclopropylpyrimidine 95%

Supplier: Ambeed
Warning

A135175-10G A135175-250MG A135175-1G A135175-5G
103802-104EA 39 USD
103802-104 76972-300 76972-302 77298-758 77298-760
4,6-Dichloro-2-cyclopropylpyrimidine 95%
4,6-Dichloro-2-cyclopropylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂
Boiling Pt: 268.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11046809
CAS Number: 7043-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR