4,6-Dichloro-2,5-dimethylpyrimidine 96%
Supplier: Ambeed
|
Warning
|
A193751-1G
A193751-5G
A193751-250MG
103816-578EA
20
USD
103816-578
103847-190
77085-092
77085-094
77287-522
4,6-Dichloro-2,5-dimethylpyrimidine 96%
4,6-Dichloro-2,5-dimethylpyrimidine
Precaución: for Research Use Only
Formula:
C₆H₆Cl₂N₂ Boiling Pt: 232.3 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09991674 CAS Number: 1780-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...