4,6-Dichloro-2,3-dihydro-1H-inden-1-one 97%
Supplier: Ambeed
|
Warning
|
A101746-100MG
A101746-250MG
A101746-1G
A101746-5G
A101746-10G
103796-112EA
15
USD
103796-112
103821-798
76821-254
76821-256
77282-818
77282-820
77610-754
4,6-Dichloro-2,3-dihydro-1H-inden-1-one 97%
4,6-Dichloro-2,3-dihydro-1H-inden-1-one
Precaución: for Research Use Only
Formula:
C₉H₆Cl₂O MW: 201.04 g/mol Boiling Pt: 326 °C (760 mm Hg) Melting Pt: 118…123 °C Flash Pt: 137 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276785 CAS Number: 52397-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...