4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A144896-100G A144896-10G A144896-250MG A144896-25G A144896-5G A144896-1G
103803-468EA 11 USD
103803-468 103841-546 77095-562 77095-564 77095-566 77299-426 77299-428 77316-804
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde 97%
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂OS
MW: 223.08 g/mol
Boiling Pt: 327.7 °C (760 mm Hg)
Melting Pt: 82…84 °C
Flash Pt: 152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00194954
CAS Number: 33097-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR